Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium triethylborohydride, 1M solution in THF, AcroSeal™, Thermo Scientific™

Catalog No. AC200041000
Click to view available options
Quantity:
100 mL

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22560-21-0, 109-99-9
Molecular Formula C6H15BK
Molecular Weight (g/mol) 137.094
MDL Number MFCD00011702
InChI Key JSWZHFBULBZRJI-UHFFFAOYSA-N
Synonym potassium triethylborohydride solution, potassium hydrotriethylborate, potassium triethylhydridoborate, potassium triethyl-? 2-boranuide, potassium triethylborohydride, 1.0m in thf, in sure/seal tm bottle
PubChem CID 23692628
IUPAC Name potassium;triethylboron(1-)
SMILES [B-](CC)(CC)CC.[K+]

Specifications

Color Undesignated
Linear Formula KB(C2H5)3H
Concentration 0.98 to 1.02M
Density 0.902g/mL
Formula Weight 138.1
Quantity 100 mL
Physical Form Solution
Chemical Name or Material Potassium triethylborohydride, 1M Solution in Tetrahydrofuran
Hazard Category Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage. May cause respiratory irritation. In contact with water releases flammable gases which may ignite spontaneously. Highly flammable liquid and vapour. Reacts violently with water. May form explosive peroxides. Suspected of causing cancer. Harmful if swallowed. May cause drowsiness or dizziness.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Handle under inert gas. Protect from moisture. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
EINECSNumber 245-077-3
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.