Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium trioxalatoferrate(III) trihydrate

Catalog No. AA3112414
Click to view available options
Quantity:
5 g
25 g
100 g
500 g

CAS: 5936-11-8 | C6H3FeK3O12 | 440.22 g/mol

Potassium trioxalatoferrate(III) trihydrate is used in chemical actinometry to measure light flux.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5936-11-8
Molecular Formula C6H3FeK3O12
Molecular Weight (g/mol) 440.22
MDL Number MFCD00150450
InChI Key YXTWCYYDGRBUDE-UHFFFAOYSA-K
Synonym Potassium iron(III) oxalate
PubChem CID 131675837

Specifications

Color Green
Physical Form Crystalline
Quantity 25 g
UN Number UN3288
Solubility Information Slightly soluble in water.
SMILES C(=O)(C(=O)O)O.C(=O)(C(=O)[O-])O.C(=O)(C(=O)[O-])[O-].[K+].[K+].[K+].[Fe]
IUPAC Name tripotassium;2-hydroxy-2-oxoacetate;iron;oxalate;oxalic acid
Formula Weight 491.26 (437.21 Anhydrous)
Sensitivity Hygroscopic
Density 2.133 g/mL
Chemical Name or Material Potassium trioxalatoferrate(III) trihydrate
Show More Show Less
Hazard Category H302+H312
Hazard Statement GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
EINECSNumber 238-954-7
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.