Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium tungsten oxide, 99.5% (metals basis)

Catalog No. p-7049334
Click to view available options
Quantity:
25 g
100 g

CAS: 7790-60-5 | K2O4W | 326.033 g/mol

Potassium tungsten oxide is used in solar energy and water treatment applications. It is used as alkaloids precipitating agent, bronze production and in tungsten refining. It is also used as an analytical reagent. Further, it is added to molten carbonate fuel cell to prevent the corrosion of stainless-steel separator and retards the sintering of the anode.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-60-5
Molecular Formula K2O4W
Molecular Weight (g/mol) 326.033
MDL Number MFCD00049668
InChI Key AAQNGTNRWPXMPB-UHFFFAOYSA-N
Synonym potassium tungstate, unii-t6vrm0ou6v, t6vrm0ou6v, dipotassium dioxido dioxo tungsten, dipotassium wolframate, potassium tungstate vi, tungstate wo42-, dipotassium, t-4, 2k.wo4, potassium tungstate-w
PubChem CID 3084032
IUPAC Name dipotassium;dioxido(dioxo)tungsten
SMILES [O-][W](=O)(=O)[O-].[K+].[K+]

Specifications

Melting Point 921°C
Physical Form Powder
Quantity 25 g
Merck Index 14,7694
Solubility Information Soluble in water.
Formula Weight 326.04
Percent Purity 99.5%
Odor Odorless
Sensitivity Hygroscopic
Density 3.12 g/mL
Assay (metals basis)
Chemical Name or Material Potassium tungsten oxide
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H319-H335
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 232-215-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.