Learn More
Invitrogen™ POTEB Polyclonal Antibody

Description
POTEB Polyclonal Antibody for Western Blot, IHC (P), Flow
Specifications
Specifications
| Antigen | POTEB |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | POTEB |
| Gene Accession No. | A0A0A6YYL3 |
| Gene Alias | A26B1; ANKRD26-like family B member 1; ANKRD26-like family B, member 1; cancer/testis antigen family 104, member 5; CT104.5; POTE ankyrin domain family member B; POTE ankyrin domain family member B2; POTE ankyrin domain family member B-like; POTE ankyrin domain family, member B; POTE15; POTE-15; POTEB; POTEB2; Prostate, ovary, testis-expressed protein on chromosome 15; protein expressed in prostate, ovary, testis, and placenta 15 |
| Gene Symbols | POTEB |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.