Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POTEB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101779
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101779 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101779 Supplier Invitrogen™ Supplier No. PA5101779
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total A26B1.

POTEB (POTE Ankyrin Domain Family Member B) is a Protein Coding gene. An important paralog of this gene is POTEB2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POTEB
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene POTEB
Gene Accession No. A0A0A6YYL3
Gene Alias A26B1; ANKRD26-like family B member 1; ANKRD26-like family B, member 1; cancer/testis antigen family 104, member 5; CT104.5; POTE ankyrin domain family member B; POTE ankyrin domain family member B2; POTE ankyrin domain family member B-like; POTE ankyrin domain family, member B; POTE15; POTE-15; POTEB; POTEB2; Prostate, ovary, testis-expressed protein on chromosome 15; protein expressed in prostate, ovary, testis, and placenta 15
Gene Symbols POTEB
Host Species Rabbit
Immunogen A synthesized peptide derived from human POTEB3(Accession A0JP26), corresponding to amino acid residues I488-L538.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100996331
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.