Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POTED Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA511495
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA511495 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA511495 Supplier Invitrogen™ Supplier No. PA511495
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

POTED Polyclonal Antibody for Western Blot, IHC (P)

The POTE protein contains seven ankyrin repeats between amino acids 140 and 380. Expression of POTE in prostate cancer and its undetectable expression in normal essential tissues make POTE a candidate for the immunotherapy of prostate cancer. The existence of a large number of closely related but rapidly diverging members, their location on multiple chromosomes and their limited expression pattern suggest an important role for the POTE gene family in reproductive processes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POTED
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene POTED
Gene Accession No. Q86YR6
Gene Alias A26B3; ANKRD21; ANKRD26-like family B member 3; ANKRD26-like family B, member 3; ankyrin repeat domain 21; ankyrin repeat domain-containing protein 21; cancer/testis antigen family 104, member 1; CT104.1; Expressed in prostate, ovary, testis, and placenta; LOC100288966; POTE; POTE ankyrin domain family member D; POTE ankyrin domain family member D-like; POTE ankyrin domain family, member D; POTE21; POTE-21; POTED; Prostate, ovary, testis-expressed protein; Protein POTE; uncharacterized protein LOC100288966
Gene Symbols LOC100288966, POTED
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 542-571 amino acids from the C-terminal region of human POTE.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100288966, 317754
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.