Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POTEE Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560301
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560301 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560301 Supplier Invitrogen™ Supplier No. PA560301
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LKALQLNELT MDDDTAVLVI DNGS Highest antigen sequence identity to the following orthologs: Mouse - 46%, Rat - 46%.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POTEE
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene POTEE
Gene Accession No. Q6S8J3
Gene Alias A26C1; A26C1A; ANKRD26-like family C member 1A; ANKRD26-like family C, member 1A; cancer/testis antigen family 104, member 2; CT104.2; POTE ankyrin domain family member E; POTE ankyrin domain family, member E; POTE2; POTE-2; POTE2gamma; POTEE; Prostate, ovary, testis-expressed protein on chromosome 2; protein expressed in prostate, ovary, testis, and placenta 2
Gene Symbols POTEE
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human POTEE. Recombinant protein control fragment (Product #RP-104357).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 445582
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.