Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POTEE Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121460
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121460 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121460 Supplier Invitrogen™ Supplier No. PA5121460
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody tested in Indirect ELISA.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POTEE
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.38 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene POTEE
Gene Accession No. Q6S8J3
Gene Alias A26C1; A26C1A; ANKRD26-like family C member 1A; ANKRD26-like family C, member 1A; cancer/testis antigen family 104, member 2; CT104.2; POTE ankyrin domain family member E; POTE ankyrin domain family, member E; POTE2; POTE-2; POTE2gamma; POTEE; Prostate, ovary, testis-expressed protein on chromosome 2; protein expressed in prostate, ovary, testis, and placenta 2
Gene Symbols POTEE
Host Species Rabbit
Immunogen Recombinant Human POTE ankyrin domain family member E protein (435-606aa).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 445582
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.