Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ POTEF Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA524128
Description
POTEF Polyclonal Antibody for Western Blot
The function of this protein remains unknown.
Specifications
POTEF | |
Polyclonal | |
Unconjugated | |
POTEF | |
A26C1B; ACTB; ANKRD26-like family C member 1B; ANKRD26-like family C, member 1B; beta actin; chimeric POTE-actin protein; POTE ankyrin domain family member F; POTE ankyrin domain family, member F; POTE-2 alpha-actin; POTE2alpha; POTEACTIN; POTEF; prostate, ovary, testis expressed protein on chromosome 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
728378 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.48 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
A5A3E0 | |
POTEF | |
KLH conjugated synthetic peptide between 656-685 amino acids from the central region of human POTEF. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction