Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
POTEG Antibody (OTI2G1), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273540B
Description
POTEG Monoclonal specifically detects POTEG in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
POTEG | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
POTEG | |
Full length recombinant protein of human POTEG (NP_001005356) produced in HEK293T cells. | |
0.1 mL | |
404785.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI2G1 | |
Western Blot, Immunohistochemistry | |
A26C2, ANKRD26-like family C, member 2, cancer/testis antigen family 104, member 4, CT104.4, POTE ankyrin domain family member G, POTE ankyrin domain family, member G, POTE14ACTBL1, POTE14alpha, POTE-14ANKRD26-like family C member 2, POTE22, Prostate, ovary, testis-expressed protein on chromosome 14, protein expressed in prostate, ovary, testis, and placenta 14 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction