Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ A26C2/A26C3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599693
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599693 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599693 Supplier Invitrogen™ Supplier No. PA599693
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total POTE-14/22.

Belongs to the POTE family. 3 isoforms of the human protein are produced by alternative splicing. Bera T.K., Gene 337:45-53(2004).
TRUSTED_SUSTAINABILITY

Specifications

Antigen A26C2/A26C3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene POTEG
Gene Accession No. Q6S545, Q6S5H5
Gene Alias 4921537P18Rik; 4930456F22Rik; A26C2; A26C3; ACTBL1; actin, beta-like 1; ANKRD26-like family C member 2; ANKRD26-like family C member 3; ANKRD26-like family C, member 2; ANKRD26-like family C, member 3; cancer/testis antigen family 104, member 4; cancer/testis antigen family 104, member 7; CT104.4; CT104.7; gonad specific ankyrin repeat protein; Gsarp1; Gsarp2; POTE ankyrin domain family member A; POTE ankyrin domain family member G; POTE ankyrin domain family member H; POTE ankyrin domain family, member G; POTE ankyrin domain family, member H; POTE14; POTE-14; POTE14alpha; POTE14B; POTE22; POTE-22; Poteg; POTEH; Prostate, ovary, testis-expressed protein on chromosome 14; prostate, ovary, testis-expressed protein on chromosome 22; protein expressed in prostate, ovary, testis, and placenta 14; protein expressed in prostate, ovary, testis, and placenta 22; protein expressed in prostate, ovary, testis, and placenta POTE14 like
Gene Symbols POTEG, POTEH
Host Species Rabbit
Immunogen A synthesized peptide derived from human POTEH(Accession Q6S545), corresponding to amino acid residues G479-E529.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23784, 404785, 70952
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.