Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POTEM Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116372
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116372 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116372 Supplier Invitrogen™ Supplier No. PA5116372
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

POTEM Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen POTEM
Applications Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene POTEM
Gene Accession No. A6NI47
Gene Alias ACT; ANKRD26-like family C member ENSP00000349402; P704P; POTE ankyrin domain family member M; POTE ankyrin domain family, member M; POTE14beta; POTEM; prostate-specific P704P; putative POTE ankyrin domain family member M
Gene Symbols POTEM
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 430-462 amino acids from the C-terminal region of human POTEM.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 641455
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.