Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
POU2AF1 Mouse anti-Human, Clone: 3C5A7, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
| Antigen | POU2AF1 |
|---|---|
| Clone | 3C5A7 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-7077
![]()
|
Proteintech Group Inc
666591IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Transcriptional coactivator that specifically associates with either OCT1 or OCT2. It boosts the OCT1 mediated promoter activity and to a lesser extent, that of OCT2. It has no intrinsic DNA-binding activity. It recognizes the POU domains of OCT1 and OCT2. It is essential for the response of B-cells to antigens and required for the formation of germinal centers.Specifications
| POU2AF1 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| BOB 1, BOB1, OBF 1, OBF1, OCA B, OCAB, OCT binding factor 1, POU2AF1 | |
| POU2AF1 | |
| IgG1 | |
| Protein G | |
| Antibody |
| 3C5A7 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q16633 | |
| 5450 | |
| POU2AF1 Fusion Protein Ag23613 | |
| Primary | |
| -20°C | |
| POU2AF1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title