Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POU2AF1 Mouse anti-Human, Clone: OTI2D4, lyophilized, TrueMAB™
SDP

Catalog No. 501668566
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-166-8566 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-166-8566 Supplier OriGene Supplier No. CF807917
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

B cell Oct binding protein 1 (BOB-1), is a transcription factor that is required for germinal center formation and immunoglobulin production. BOB-1 is expressed at high levels in spleen and peripheral blood leukocytes and represents an OCT co-factor capable of conferring cell specific activation of OCT-1 and OCT-2.

Specifications

Antigen POU2AF1
Applications Western Blot
Classification Monoclonal
Clone OTI2D4
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene POU2AF1
Gene Accession No. Q16633
Gene Alias BOB1, OBF-1, OBF1, OCAB
Gene Symbols POU2AF1
Host Species Mouse
Immunogen Full length human recombit protein of human POU2AF1 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5450
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.