Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POU3F2 Monoclonal Antibody (6H12)
GREENER_CHOICE

Catalog No. PIMA517153
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA517153 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA517153 Supplier Invitrogen™ Supplier No. MA517153
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-17153 targets POU3F2 in indirect ELISA and WB applications and shows reactivity with Human samples. The MA5-17153 immunogen is purified recombinant fragment of human POU3F2 (amino acids: 214-362) expressed in E. Coli. MA5-17153 detects POU3F2 which has a predicted molecular weight of approximately 46.9kDa.

This gene encodes a member of the POU-III class of neural transcription factors. The encoded protein is involved in neuronal differentiation and enhances the activation of corticotropin-releasing hormone regulated genes. Overexpression of this protein is associated with an increase in the proliferation of melanoma cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POU3F2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 6H12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene POU3F2
Gene Accession No. P20265
Gene Alias 7-Oct; 9430075J19Rik; A230098E07Rik; Brain-2; brain-2 class III POU-domain protein; brain-specific homeobox/POU domain protein 2; BRN2; Brn-2; Nervous system-specific octamer-binding transcription factor N-Oct-3; N-Oct3; OCT7; Oct-7; Octamer-binding protein 7; octamer-binding transcription factor 7; Otf7; OTF-7; POU class 3 homeobox 2; POU domain, class 3, transcription factor 2; POU3F2; POUF3; protein Brn-2; Unknown (protein for MGC:137739)
Gene Symbols POU3F2
Host Species Mouse
Immunogen Purified recombinant fragment of human POU3F2 (amino acids: 214-362) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5454
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.