Learn More
Invitrogen™ POU3F2 Polyclonal Antibody

Description
PA5-23400 targets POU3F2 in IHC (P) and WB applications and shows reactivity with Bovine, Canine, Equine, Human, mouse, and Rat samples. The PA5-23400 immunogen is synthetic peptide (254 AHHDPHSDEDTPTSDD 269) of POU3F2 protein. The amino acid sequence used as immunogen is 87% homologous in Equine, 81% homologous in Bovine and Porcine.
Specifications
Specifications
| Antigen | POU3F2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA and 0.05% sodium azide |
| Gene | POU3F2 |
| Gene Accession No. | P20265, P31360, P56222 |
| Gene Alias | 7-Oct; 9430075J19Rik; A230098E07Rik; Brain-2; brain-2 class III POU-domain protein; brain-specific homeobox/POU domain protein 2; BRN2; Brn-2; Nervous system-specific octamer-binding transcription factor N-Oct-3; N-Oct3; OCT7; Oct-7; Octamer-binding protein 7; octamer-binding transcription factor 7; Otf7; OTF-7; POU class 3 homeobox 2; POU domain, class 3, transcription factor 2; POU3F2; POUF3; protein Brn-2; Unknown (protein for MGC:137739) |
| Gene Symbols | POU3F2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.