Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POU3F2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534469
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534469 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534469 Supplier Invitrogen™ Supplier No. PA534469
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is NIH-3T3 cell lysate. PA5-34469 can be used with blocking peptide PEP-1511.

This gene encodes a member of the POU-III class of neural transcription factors. The encoded protein is involved in neuronal differentiation and enhances the activation of corticotropin-releasing hormone regulated genes. Overexpression of this protein is associated with an increase in the proliferation of melanoma cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POU3F2
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene POU3F2
Gene Accession No. P20265, P31360, P56222
Gene Alias 7-Oct; 9430075J19Rik; A230098E07Rik; Brain-2; brain-2 class III POU-domain protein; brain-specific homeobox/POU domain protein 2; BRN2; Brn-2; Nervous system-specific octamer-binding transcription factor N-Oct-3; N-Oct3; OCT7; Oct-7; Octamer-binding protein 7; octamer-binding transcription factor 7; Otf7; OTF-7; POU class 3 homeobox 2; POU domain, class 3, transcription factor 2; POU3F2; POUF3; protein Brn-2; Unknown (protein for MGC:137739)
Gene Symbols POU3F2
Host Species Rabbit
Immunogen An 18 amino acid peptide near the carboxy terminus of human POU3F2.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18992, 29588, 5454
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.