Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POU3F3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA519305
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA519305 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA519305 Supplier Invitrogen™ Supplier No. PA519305
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with human based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 4,000.

POU3F3 is a member of the class III POU family of transcription factors that are expressed in the central nervous system. The POU domain in these proteins is required for high affinity binding to octamer DNA sequences.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POU3F3
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene POU3F3
Gene Accession No. P31361
Gene Alias 8-Oct; Brain-1; brain-specific homeobox/POU domain protein 1; Brn1; Brn-1; brn-1 protein; class III POU protein POU-domain; oct-8; Octamer-binding protein 8; octamer-binding transcription factor 8; Otf8; OTF-8; POU class 3 homeobox 3; POU domain, class 3, transcription factor 3; Pou3f3; RHS1; Rhs2; Skin1
Gene Symbols POU3F3
Host Species Goat
Immunogen Synthetic peptide sequence (HMLSHAHQWVTAL) corresponding to the internal amino acids of POU3F3 (aa 88-101).
Purification Method Antigen affinity chromatography, Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18993
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.