Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
POU3F3 Polyclonal Antibody
POU3F3 Polyclonal Antibody
Supplier: Thermo Scientific PA565740
Description
POU3F3 Polyclonal Antibody for ICC/IF

Specifications
POU3F3 | |
Polyclonal | |
Unconjugated | |
POU3F3 | |
8-Oct; Brain-1; brain-specific homeobox/POU domain protein 1; Brn1; Brn-1; brn-1 protein; class III POU protein POU-domain; oct-8; Octamer-binding protein 8; octamer-binding transcription factor 8; Otf8; OTF-8; POU class 3 homeobox 3; POU domain, class 3, transcription factor 3; Pou3f3; RHS1; Rhs2; Skin1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5455 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P20264 | |
POU3F3 | |
Recombinant Human POU3F3. Recombinant protein control fragment (Product #RP-104002). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction