Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ POU3F3A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572619
Description
POU3F3A Polyclonal Antibody for Western Blot
Transcription factor that may play important roles in patterning the embryonic brain.
Specifications
POU3F3A | |
Polyclonal | |
Unconjugated | |
pou3f3a | |
alternatively spliced brn-1.1 protein variant; brain POU domain gene 1.1; brain-1.1; Brain-11; Brain-specific homeobox/POU domain protein 1.1; brn1.1; brn-1.1; brn11; brn-11; class III POU domain protein taichi; POU class 3 homeobox 3a; POU domain gene 12; POU domain protein; POU domain protein 12; POU domain protein zp-12; POU domain, class 3, transcription factor 3-A; pou12; pou12 protein; pou3f3a; si:dkeyp-118h3.2; tai-ji; type III POU domain protein; wu:fc33a11; zfbrn-1.1; zfBrn-11; zp12; ZP-12; zp12pou | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
30386 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.95 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P56224 | |
pou3f3a | |
KLH conjugated synthetic peptide between 381-407 amino acids from the C-terminal region of DANRE pou3f3a. | |
400 μL | |
Primary | |
Human, Zebrafish | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction