Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POU3F4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556895
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556895 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556895 Supplier Invitrogen™ Supplier No. PA556895
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RSPHVAHHSP HTNHPNAWGA SPAPNPSITS SGQPLNVYSQ PGFTVSGMLE HGGLTPPPAA ASAQSLHPVL REPPDHGELG SHHCQDH Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

POU3F4 (brain-specific homeobox/POU domain protein 4, brain-4, Brn-4, transcription factor 4) is a transcription factor with a POU domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POU3F4
Applications Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene POU3F4
Gene Accession No. P49335
Gene Alias brain-4; brain-4 class III POU-domain protein; brain-specific homeobox/POU domain protein 4; BRN4; Brn-4; DFN3; DFNX2; OCT-9; Octamer-binding protein 9; octamer-binding transcription factor 9; Otf9; OTF-9; POU class 3 homeobox 4; POU domain, class 3, transcription factor 4; POU3F4; RHS2; RHS2 class III POU protein; sex linked fidget; Slf
Gene Symbols POU3F4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human POU3F4. Recombinant protein control fragment (Product #RP-95728).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5456
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.