Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
POU4F3 Antibody (OTI4C5), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273541PCP
Description
POU4F3 Monoclonal antibody specifically detects POU4F3 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
POU4F3 | |
Monoclonal | |
PerCP | |
PBS | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
OTI4C5 | |
Western Blot | |
brain-3C, Brain-specific homeobox/POU domain protein 3C, BRN3.1, Brn-3C, BRN3Cbrn-3C, DFNA15, MGC138412, POU class 4 homeobox 3, POU domain class 4, transcription factor 3, POU domain, class 4, transcription factor 3 | |
Human recombinant protein fragment corresponding to amino acids 179-338 of human POU4F3(NP_002691) produced in E.coli. | |
0.1 mL | |
Neuroscience, Vision | |
5459 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction