Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POU4F3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596769
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596769 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596769 Supplier Invitrogen™ Supplier No. PA596769
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MMAMNSKQPF GMHPVLQEPK FSSLHSGSEA MRRVCLPAPQ LQGNIFGSFD ESLLARAEAL AAVDIVSHGK NHPFKPDATY HTMSSVPCTS TSSTVPISHP AALTSHPHHA VHQGLEGDLL EHISPTLSVS GLGAPEHSVM PAQIHPHHLG AMGHLHQAMG MSHPHTVAPH SAMPACLSDV; Positive Samples: Jurkat; Cellular Location: Nucleus.

POU4F3 encodes a member of the POU-domain family of transcription factors. POU-domain proteins have been observed to play important roles in control of cell identity in several systems. This protein is found in the retina and may play a role in determining or maintaining the identities of a small subset of visual system neurons. Defects in this gene are the cause of non-syndromic sensorineural deafness autosomal dominant type 15.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POU4F3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.88 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Pou4f3
Gene Accession No. Q15319
Gene Alias brain POU domain gene 3.1; brain-3.1; Brain-3C; brain-specific homeobox POU domain protein 3C; brain-specific homeobox/POU domain protein 3.1; Brain-specific homeobox/POU domain protein 3C; brn3.1; Brn-3.1; brn3c; brn-3c; ddl; DFNA15; dreidel; MGC138412; POU class 4 homeobox 3; POU domain transcription factor Brn3C; POU domain, class 4, transcription factor 3; pou4f3; zfbrn-3.1
Gene Symbols Pou4f3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-180 of human POU4F3 (NP_0026911).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5459
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.