Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POU5F1 Mouse anti-Human, Clone: 9B7, Abnova™

Catalog No. 89035526 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-526 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-526 Supplier Abnova Corporation Supplier No. MAB14128

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against full length recombinant human POU5F1.

  • High specificity
  • High reactivity

Specifications

Antigen POU5F1
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 9B7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant human POU5F1.
Dilution Flow Cytometry (3-5 ug x 106 cells) Immunocytochemistry (1-2 ug/mL) Immunofluorescence (1-2 ug/mL) Western Blot (1:500-1:5000) The optimal working dilution should be determined by the end user.
Formulation In PBS (30% glycerol, 1 mg/mL BSA, 0.05% sodium azide).
Gene Accession No. Q01860
Gene Alias MGC22487/OCT3/OCT4/OTF3/OTF4
Gene Symbols POU5F1
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human POU5F1.
Purification Method Protein A affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5460
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.