Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POU5F1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566155
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566155 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566155 Supplier Invitrogen™ Supplier No. PA566155
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VGPGSEVWGIP PCPPPYELCG GMAYCGPQVG VGLVPQGGLE TSQPESEAGV Highest antigen sequence identity to the following orthologs - mouse 76%, rat 76%.

This intronless gene was thought to be a transcribed pseudogene of POU class 5 homeobox 1, however, it has been reported that this gene can encode a functional protein. The encoded protein is nearly the same length as and highly similar to the POU class 5 homeobox 1 transcription factor, has been shown to be a weak transcriptional activator and may play a role in carcinogenesis and eye development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POU5F1B
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene POU5F1B
Gene Accession No. Q06416
Gene Alias OCT4PG1; Oct4-pg1; octamer binding protein 3_like sequence; Octamer-binding protein 3-like; Octamer-binding transcription factor 3-like; OTF3C; OTF3P1; POU 5 domain protein; POU class 5 homeobox 1 pseudogene 1; POU class 5 homeobox 1B; POU domain transcription factor Oct-4; POU domain transcription factor OCT4-pg1; POU domain, class 5, transcription factor 1 pseudogene 1; POU5F1B; POU5F1P1; POU5F1P4; POU5FLC20; POU5FLC8; Putative POU domain, class 5, transcription factor 1B
Gene Symbols POU5F1B
Host Species Rabbit
Immunogen Recombinant Human POU5F1B. Recombinant protein control fragment (Product #RP-105098).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5462
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.