Learn More
Invitrogen™ POU5F2 Monoclonal Antibody (3E3)

Description
Peptide Sequence: AGHRPSNHFC PLPGSGGGGP RGPMPLRVDT LTWLSTQAAP GRVMVWPAVR PGICPGPDVW RIPLGPLPHE FRGWIAPCRP RLGASEAGDW LRRPSEGAL.
Specifications
Specifications
| Antigen | POU5F2 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 3E3 |
| Concentration | 0.2-1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | POU5F2 |
| Gene Accession No. | Q8N7G0 |
| Gene Alias | 1700013G10Rik; POU domain class 5, transcription factor 2; POU domain, class 5, transcription factor 2; Pou5f2; Sperm 1 POU domain transcription factor; sperm-associated POU domain protein 1; Sprm1; SPRM-1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.