Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POU5F2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5139955
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5139955 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5139955 Supplier Invitrogen™ Supplier No. PA5139955
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: QQLSVANMWK LRPLLKKWLK EVEAENLLGL CKMEMILQQS GKWRRASRER RIGNSLEKFF QRCPKP Highest antigen sequence indentity to the following orthologs: Rat - 74%, Mouse - 73%.

Transcription factor that binds preferentially to the octamer motif (5'-ATGTTAAT-3'). May exert a regulatory function in meiotic events that are required for terminal differentiation of male germ cell.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POU5F2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene POU5F2
Gene Accession No. Q8N7G0
Gene Alias 1700013G10Rik; POU domain class 5, transcription factor 2; POU domain, class 5, transcription factor 2; Pou5f2; Sperm 1 POU domain transcription factor; sperm-associated POU domain protein 1; Sprm1; SPRM-1
Gene Symbols POU5F2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human POU5F2. Recombinant protein control fragment (Product #RP-109638).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 134187
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.