Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POU5F2 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501735317 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5317 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5317 Supplier Proteintech Group Inc Supplier No. 263621AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Transcription factor that binds preferentially to the octamer motif (5′-ATGTTAAT-3′). May exert a regulatory function in meiotic events that are required for terminal differentiation of male germ cell.

Specifications

Antigen POU5F2
Applications Western Blot
Classification Polyclonal
Concentration 0.37 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene POU5F2
Gene Accession No. Q8N7G0
Gene Alias POU5F2, SPRM 1, SPRM1
Gene Symbols POU5F2
Host Species Rabbit
Immunogen POU5F2 Fusion Protein Ag23803
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 134187
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.