Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POU6F1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565617
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565617 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565617 Supplier Invitrogen™ Supplier No. PA565617
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KPSASAPIPIT CSETPTVSQL VSKPHTPSLD EDGINLEEI Highest antigen sequence identity to the following orthologs - mouse 95%, rat 100%.

Brn-5 (Brain-5), also known as POU6F1, MPOU or TCFB1, is a widely expressed protein but during embryogenesis is exclusively found in the developing brain and spinal cord. As is characteristic of Brn family members, Brn-5 contains two DNA-binding domains, namely the POU-specific domain and the POU homeodomain, which each contain an HTH (helix-turn-helix) motif. Brn-5 binds to CRH (corticotrophin-releasing hormone) elements with high affinity and is capable of both enhancing Prolactin gene expression and activating Pit-1 expression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POU6F1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene POU6F1
Gene Accession No. Q14863
Gene Alias 2310038G18Rik; brain-5; Brain-specific homeobox/POU domain protein 5; BRN5; brn-5; brn-5 protein; C79593; cns-1; Emb; MPOU; mPOU homeobox protein; Octamer-binding transcription factor EMB; POU class 6 homeobox 1; POU domain, class 6, transcription factor 1; Pou6f1; TCFB1; transcription regulatory protein MCP-1
Gene Symbols POU6F1
Host Species Rabbit
Immunogen Recombinant Human POU6F1. Recombinant protein control fragment (Product #RP-106284).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5463
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.