Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POU6F2 Monoclonal Antibody (8F9)
GREENER_CHOICE

Catalog No. PIMA521327
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA521327 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA521327 Supplier Invitrogen™ Supplier No. MA521327
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: IAGQVSKPLL SVRSEMNAEL RGEDKAATSD SELNEPLLAP VESNDSEDTP SKLFGARGNP ALSDPGTPDQ HQASQTHPPF PVGPQP.

This gene encodes a member of the POU protein family characterized by the presence of a bipartite DNA binding domain, consisting of a POU-specific domain and a homeodomain, separated by a variable polylinker. The DNA binding domain may bind to DNA as monomers or as homo- and/or heterodimers, in a sequence-specific manner. The POU family members are transcriptional regulators, many of which are known to control cell type-specific differentiation pathways. This gene is a tumor suppressor involved in Wilms tumor (WT) predisposition. Alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POU6F2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 8F9
Concentration 0.2-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Pou6f2
Gene Accession No. P78424
Gene Alias D130006K24Rik; POU class 6 homeobox 2; POU domain class 6 transcription factor 2; POU domain, class 6, transcription factor 2; Pou6f2; Retina-derived POU domain factor 1; retina-derived POU-domain factor-1; RPF1; RPF-1; Wilms tumor suppressor locus; WT5; WTSL
Gene Symbols Pou6f2
Host Species Mouse
Immunogen POU6F2 (NP_009183, 2 a.a. ~ 87 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11281
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.