Learn More
Invitrogen™ Powassan Virus Monoclonal Antibody (3683)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA547518
Description
Specificity: Specific for Powassan virus. Negative vs. related viruses. Nonreactive with: West Nile virus, Eastern Equine Encephalitis virus, Western Equine Encephalitis virus, St. Louis Encephalitis virus and La Crosse virus. Purified preparations consist of >90% pure mouse monoclonal antibody which has been purified from ascites fluid by protein A chromatography. The final preparation is formulated to a protein concentration of 100 μg/mL in 0.01 M phosphate buffered saline, pH 7.2 and contains 0.1% sodium azide. Each vial contains 1.0 mL. This product contains no stabilizing proteins and should be stored at 4C until ready for use. Working dilution must be determined by the user. Suggested starting ranges are 1:10-1:50 for ICC/IF and 1:20-1:200. for ELISA.
Powassan virus is a tickborne flavivirus.
Specifications
Powassan Virus | |
Monoclonal | |
100 μg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Proprietary sequence. | |
100 μg | |
Primary | |
4°C | |
Liquid |
ELISA, Immunocytochemistry | |
3683 | |
Unconjugated | |
Mouse | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG2b |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.