Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Power Hawk AUTO CRIB-IT™ Automatic Stabilization Tool

Catalog No. 19166797 Shop All Power Hawk Technologies, Inc. Products
Encompass
Change view
Click to view available options
Material:
Aluminum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-166-797 Aluminum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-166-797 Supplier Power Hawk Supplier No. AC200
Only null left
Add to Cart
Add to Cart

Spring-activated for vehicle stabilization in seconds

Super-compact, high-strength aluminum device weighs only 14.5 lbs. but is rated for 2000 lb. maximum load.

  • AUTO CRIB-IT is taller than most step chocks—up to 13 in. in height
  • Automatically re-adjusts and locks during operations
  • Features quick and easy quarter-turn lock
  • Can be positioned completely under and parallel to rocker panels to avoid trip hazards
  • Easy to decontaminate

Order Info

Storage Dimensions: 28L x 4W x 2.75 in.; Sold as set of two.

Specifications

Product Type Stabilization Device
Material Aluminum
Height (English) 13 in.
Height (Metric) 33.02 cm
Load Capacity (English) 2000 lb.
Load Capacity (Metric) 907 kg
Weight (English) 15 lb.
Weight (Metric) 6.8 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.