Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZOLL™ Powerheart™ G5/G3 AED Rescue Backpack
Shop All Zoll Medical Corporation Products
Click to view available options
Product Type:
Rescue Backpack
Description
- Easily accessible internal compartment
- Space for rescue supplies
- Pads' expiration date is visible at all times
Compatible With:
- Powerheart™ G3
- Powerheart™ G5
Specifications
Specifications
| Product Type | Rescue Backpack |
| Color | Black |
| For Use With (Application) | For medic teams, search-and-rescue and mobile first responders |
| For Use With (Equipment) | Powerheart™ G3 AED, Powerheart™ G5 AED |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction