Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ PowerTrack SYBR Green Master Mix with Choice of Fast or Standard PCR Plates Bundle PROMO

Catalog No. A48345PM
Encompass_Preferred
Change view
Click to view available options
Reaction Speed:
Standard
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Reaction Speed
A48345PM Standard
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A48345PM Supplier Applied Biosystems™ Supplier No. A48345PM
Get Up to 48% Off Gene Expression Packages
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

qPCR reagent and consumable bundle featuring PowerTrack SYBR Master Mix (2 x 5 mL), 96-well reaction plates, and optical adhesive covers for reliable, high-performance real-time PCR workflows with consistent results and efficient sample processing.

Support accurate and efficient real-time PCR workflows with this convenient bundle of optimized reagents and essential consumables. Designed for consistency and ease of use, this package combines PowerTrack™ SYBR™ Master Mix with high-quality 96-well reaction plates and optical adhesive covers to enable reliable gene expression analysis and routine qPCR applications.

By bringing together key workflow components in a single SKU, this bundle simplifies ordering, ensures compatibility, and helps laboratories maintain reproducibility across experiments.

What's Included

  • PowerTrack™ SYBR™ Master Mix (2 x 5 mL)
  • 96-well thermal cycling reaction plates
  • Optical adhesive covers for qPCR applications

Features

  • Optimized SYBR Green chemistry: PowerTrack™ SYBR™ Master Mix is formulated to deliver strong amplification performance, high sensitivity, and consistent fluorescence detection for real-time PCR assays.
  • Ready-to-use reagent format: Simplifies workflow setup and reduces pipetting steps, helping minimize variability and improve reproducibility.
  • High-quality reaction plates: 96-well plates are designed for uniform thermal transfer and compatibility with standard qPCR instruments, supporting consistent amplification across all wells.
  • Reliable plate sealing: Optical adhesive covers provide secure sealing to minimize evaporation and prevent sample loss during thermal cycling.
  • Streamlined workflow: Bundled reagents and consumables ensure compatibility and reduce the need to source components separately.
  • Versatile application support: Suitable for gene expression analysis, assay optimization, and routine real-time PCR workflows.

Benefits

  • Consistent, reproducible results with optimized SYBR Green chemistry and high-quality consumables designed to work together.
  • Improved workflow efficiency by reducing setup time and simplifying reagent preparation.
  • Reduced procurement complexity with a single bundle that includes essential qPCR components.
  • Enhanced data reliability through uniform thermal performance and secure plate sealing.
  • Flexible use across applications including gene expression studies and routine qPCR assays.

This PowerTrack™ SYBR™ qPCR bundle provides a dependable, easy-to-use solution for laboratories seeking to streamline workflows while maintaining high performance and data quality in real-time PCR experiments.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X (Master Mix)
Detection Method SYBR
Form Liquid
GC-Rich PCR Performance Medium
PCR Method Standard PCR, 2-step RT-qPCR
Reaction Speed Standard
Label or Dye SYBR Green
Product Type Master Mix
For Use With (Application) Gene Expression, siRNA Validation, Pre-amplification, DNA Quantitation
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.