Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ PowerTrack SYBR Green Master Mix with PCR Plates Bundle PROMO

Catalog No. A48344PM
Encompass_Preferred
Change view
Click to view available options
Detection Method:
SYBR
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detection Method
A48344PM SYBR
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A48344PM Supplier Applied Biosystems™ Supplier No. A48344PM
Get Up to 48% Off Gene Expression Packages
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Switch to the affordable solution that eases optimization and pipetting efforts while delivering unsurpassed specificity. Choose the fast 0.1 mL PowerTrack SYBR Green Master Mix and Optical MicroAmp Reaction Plates and Adhesive covers bundle option for faster PCR.

Fast Bundle Includes (A48344)

Includes:

  • A46110 PowerTrack SYBR Green Master Mix 2 x 5 mL
  • 4483485 MicroAmp Fast Plates - 20 Plates
  • 4311971 MicroAmp Plate Covers - 100 Covers

Features:

  • Reduce total reaction time from 2 hours to 25 minutes
  • Two-color tracking dye reduces pipetting errors
  • Broad dynamic range eases optimization efforts
  • High specificity yields confident results
  • Combine with SuperScript IV VILO Master Mix for clearly better PCR

Standard Bundle Includes (A48345)

Includes:

  • A46110 PowerTrack SYBR Green Master Mix 2 x 5 mL
  • 4306737 MicroAmp Plates - 20 Plates
  • 4311971 MicroAmp Plate Covers - 100 Covers

Features:

  • MicroAmp PCR plates deliver unmatched uniformity
  • Two-color tracking dye reduces pipetting errors
  • Broad dynamic range eases optimization efforts
  • High sensitivity yields confident results
  • Combine with SuperScript IV VILO Master Mix for clearly better PCR
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 10 x 0.1 mL Applied Biosystems™ MicroAmp Fast Optical 96-well Reaction Plate (Cat. No. 4483485), 100 x Applied Biosystems™ MicroAmp Optical Adhesive Film (Cat. No. 4311971), 2 x 5 mL PowerTrack SYBR Green Master Mix 96-well (Cat. No. A46110)
Detection Method SYBR
Reaction Speed Fast
Includes Label or Dye Yes
For Use With (Application) Real Time PCR (qPCR)
Includes MicroAmp Fast Optical 96-well Reaction Plates, 0.1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.