Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel Art™ Poxygrid™ Wire Test Tube Racks

Catalog No. 22260548
Click to view available options
Color:
Blue
Green
Orange
Yellow
Positions:
48
60
72
108
150

Chemical (organic solvents, salts, weak acids and weak alkalis) and corrosion-resistant steel wire racks are coated with heat-cured epoxy

Chemical (organic solvents, salts, weak acids and weak alkalis) and corrosion-resistant steel wire racks are coated with heat-cured epoxy

  • Withstands temperature range of -223°C (-369°F) to 121°C (250°F)
  • Consists of square holes, three decks, a slotted grid bottom and extra strength reinforced edges
  • Orange racks can be used for biohazard warning while yellow for radioactive warning
  • Epoxy coatings are unaffected by organic solvents, salts, weak acids and weak alkalis
  • Stores from 40 to 108 tubes (five different diameters of testtubes)
  • Autoclavable

Specifications

Product Type Test Tube Rack
Array 4 x 12
Autoclavable Autoclavable
Color Green
Holds Tubes (13 to 16 mm)
Material Epoxy-coated Steel
Positions 48
Shape Rectangular
Shelves 3 Tiers
Stackable Yes, When Empty
Sterility Non-sterile
Dimensions (L x W x H) 9.5 x 3.6 x 2.5 in. (242 x 92 x 64 mm)
Length (Metric) 242 mm
Length (English) 9.48 in.
Width (Metric) 92 mm
Width (English) 3.6 in.
Height (Metric) 64 mm
Height (English) 2.51 in.
Diameter (Metric) Opening 16 mm
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.