Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PP 1
GREENER_CHOICESDP

Catalog No. 139710 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-971-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 13-971-0 Supplier Tocris Bioscience™ Supplier No. 1397/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent, selective Src family kinase inhibitor

Potent inhibitor of Src-family tyrosine kinases. Inhibits p56lck and p59fynT (IC50 values are 5 and 6 nM respectively). Displays > 8000-fold selectivity over ZAP-70 and JAK2. Also moderately inhibits p38, CSK, PDGF receptors, RET-derived oncoproteins, c-Kit and Bcr-Abl.

Chemical Identifiers

CAS 172889-26-8
Molecular Formula C16H19N5

Specifications

Quantity 10 mg
Formula Weight 281.36
Purity >99%
Molecular Weight (g/mol) 281.363
Chemical Name or Material PP 1
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.