Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PP 242
GREENER_CHOICE

Catalog No. 425725
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-572-5 25 mg
42-575 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 42-572-5 Supplier Tocris Bioscience™ Supplier No. 4257/25
Add to Cart
Add to Cart
This item is not returnable. View return policy

Dual mTORC1/mTORC2 inhibitor

PP 242 is an ATP-competitive mTORC1/mTORC2 inhibitor (IC50 = 8 nM). Displays selectivity for mTOR over other PI 3K family kinases (IC50 values are 0.102, 0.408, 1.27, 1.96 and 2.2 μM for p110γ, DNA-PK, p110δ, p110α and p110β respectively) and 215 further kinases. Displays modest inhibition of PKCα, JAK2, PKCβI, PKCβII and RET (IC50 values are 0.049, 0.110, 0.185, 0.198 and 0.224 μM respectively). Inhibits both S6K and 4EBP1 phosphorylation; activity causes a decrease in cap-dependent protein translation. Also triggers downregulation of cFLIPS and augments TRAIL-induced apoptosis of cancer cells. PP 242 also inhibits oncogenic K-Ras and PI 3-K induced lipogenesis in breast cancer cells.

Specifications

Chemical Name or Material 2-[4-Amino-1-(1-methylethyl)-1H-pyrazolo[3,4-d]pyrimidin-3-yl]-1H-indol-5-ol
CAS 1092351-67-1
Assay Percent Range >98%
Quantity 25 mg
Molecular Formula C16H16N6O
Formula Weight Observed MW: 308.34
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.