Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PP 3
GREENER_CHOICESDP

Catalog No. 279410 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-941-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 27-941-0 Supplier Tocris Bioscience™ Supplier No. 2794/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Negative control for PP 2 (Cat. No. 1407)

Negative control for the Src kinase inhibitor, PP 2. Inhibits EGFR kinase (IC50 = 2.7 μM).

Active Analog also available.

Chemical Identifiers

CAS 5334-30-5
Molecular Formula C11H9N5

Specifications

Quantity 10 mg
Formula Weight 211.22
Purity >98%
Molecular Weight (g/mol) 211.23
Chemical Name or Material PP 3
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.