Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PP1 alpha/beta/gamma Recombinant Rabbit Monoclonal Antibody (HL1677)
GREENER_CHOICE

Catalog No. PIMA547138
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547138 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547138 Supplier Invitrogen™ Supplier No. MA547138
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial.

PP1 is a serine/threonine specific protein phosphatase known to be involved in the regulation of a variety of cellular processes, such as cell division, glycogen metabolism, muscle contractility, protein synthesis, and HIV-1 viral transcription Increased PP1 activity has been observed in the end stage of heart failure Studies suggest that PP1 is an important regulator of cardiac function and that PP1 deregulation is implicated in diabetes and multiple types of cancer PP1 alpha is one of the three catalytic subunits of protein phosphatase 1 (PP1) This broadly expressed gene encodes the alpha subunit of the PP1 complex that associates with over 200 regulatory proteins to form holoenzymes which dephosphorylate their biological targets with high specificity PP1 beta encodes the beta isozyme and PP1 gamma encodes the gamma isozyme.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PP1 alpha/beta/gamma
Applications Western Blot
Classification Recombinant Monoclonal
Clone HL1677
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Accession No. P29466, P48729, Q8N752
Gene Alias HEL-S-80p; NSLH2; PP1A; PP-1A; PP1alpha; PP1B; PP-1B; PP1beta; PP1C; PP-1G; PPP1A; PPP1CD; PPP1G; protein phosphatase 1 catalytic subunit alpha; protein phosphatase 1 catalytic subunit beta; protein phosphatase 1 catalytic subunit gamma
Host Species Rabbit
Immunogen Recombinant fragment of human PP1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 122011, 1452, 834
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.