Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PP1 alpha/PPP1A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $646.00
Specifications
Antigen | PP1 alpha/PPP1A |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
PP1 alpha/PPP1A Polyclonal specifically detects PP1 alpha/PPP1A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
PP1 alpha/PPP1A | |
Polyclonal | |
Rabbit | |
Protein Phosphatase | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
EC 3.1.3.16, PP1alpha, PP-1AMGC15877, PPP1AMGC1674, protein phosphatase 1, catalytic subunit, alpha isoform, protein phosphatase 1, catalytic subunit, alpha isozyme, serine/threonine protein phosphatase PP1-alpha 1 catalytic subunit, serine/threonine-protein phosphatase PP1-alpha catalytic subunit | |
PPP1CA | |
IgG | |
Affinity Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
P62136 | |
5499 | |
This antibody was developed against a recombinant protein corresponding to amino acids: QILKPADKNKGKYGQFSGLNPGGRPIT | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title