Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PP1 alpha/PPP1A Antibody (OTI6E5), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273542R
Description
PP1 alpha/PPP1A Monoclonal specifically detects PP1 alpha/PPP1A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
PP1 alpha/PPP1A | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PPP1CA | |
Full length recombinant protein of human PP1 alpha/PPP1A (NP_001008709) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI6E5 | |
Western Blot, Immunohistochemistry | |
EC 3.1.3.16, PP1alpha, PP-1AMGC15877, PPP1AMGC1674, protein phosphatase 1, catalytic subunit, alpha isoform, protein phosphatase 1, catalytic subunit, alpha isozyme, serine/threonine protein phosphatase PP1-alpha 1 catalytic subunit, serine/threonine-protein phosphatase PP1-alpha catalytic subunit | |
Mouse | |
Affinity Purified | |
Phospho Specific, Protein Phosphatase | |
5499.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction