Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PP1 beta Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102349
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102349 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102349 Supplier Invitrogen™ Supplier No. PA5102349
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PPP1CB.

The protein encoded by this gene is one of the three catalytic subunits of protein phosphatase 1 (PP1). PP1 is a serine/threonine specific protein phosphatase known to be involved in the regulation of a variety of cellular processes, such as cell division, glycogen metabolism, muscle contractility, protein synthesis, and HIV-1 viral transcription. Mouse studies suggest that PP1 functions as a suppressor of learning and memory. Two alternatively spliced transcript variants encoding distinct isoforms have been observed.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PP1 beta
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Ppp1cb
Gene Accession No. P62140, P62141, P62142
Gene Alias 1200010B19; epididymis secretory sperm binding protein Li 80p; HEL-S-80p; hypothetical protein LOC538829; MGC21657; PP1B; PP-1B; PP1beta; PP1-BETA; PP2CB; PP2CBETA; PP2C-beta-X; PPC2BETAX; PPP1CB; PPP1CD; protein phosphatase 1; protein phosphatase 1 catalytic subunit beta; protein phosphatase 1, catalytic subunit, beta isoform; protein phosphatase 1, catalytic subunit, beta isozyme; protein phosphatase 1, catalytic subunit, delta isoform; protein phosphatase 1-beta; protein phosphatase 1-delta; protein phosphatase-1a; serine/threonine protein phosphatase PP1-beta catalytic subunit; serine/threonine-protein phosphatase PP1-beta catalytic subunit
Gene Symbols Ppp1cb
Host Species Rabbit
Immunogen A synthesized peptide derived from human PPP1CB(Accession P62140), corresponding to amino acid residues S298-R327.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19046, 25594, 5500
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.