Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PP1 gamma Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119827
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119827 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5119827 Supplier Invitrogen™ Supplier No. PA5119827
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Control: 293 cell lysate, mouse lung tissue lysate, rat brain tissue lysate, human prostate tissue, human stomach tissue. Subcellular Location: Nucleus, Mitochondrion, Cytoplasm, Kinetochore, Cleavage furrow, Midbody.

The protein encoded by this gene belongs to the protein phosphatase family, PP1 subfamily. PP1 is an ubiquitous serine/threonine phosphatase that regulates many cellular processes, including cell division. It is expressed in mammalian cells as three closely related isoforms, alpha, beta/delta and gamma, which have distinct localization patterns. This gene encodes the gamma isozyme. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PP1 gamma
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene PPP1CC
Gene Accession No. P36873, P63087, P63088
Gene Alias dis2m1; MGC1675; MGC2870; PP1; PP1C; PP1C gamma 1; PP1C gamma 2; PP1G; PP-1G; PP1gamma; PP2CG; PP2CGAMMA; PPP1CC; Ppp1cc1; PPP1G; PPP2CG; protein phosphatase 1 catalytic subunit gamma; protein phosphatase 1, catalytic subunit, gamma isoform; protein phosphatase 1, catalytic subunit, gamma isozyme; protein phosphatase 1C catalytic subunit; protein serine-threonine phosphatase catalytic subunit PP-1b; serine/threonine phosphatase 1 gamma; serine/threonine-protein phosphatase PP1-gamma catalytic subunit
Gene Symbols PPP1CC
Host Species Rabbit
Immunogen Synthetic peptide within human PP1C gamma aa 293-323.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19047, 24669, 5501
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.