Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PP1 gamma Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112122
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112122 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112122 Supplier Invitrogen™ Supplier No. PA5112122
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody reacts with: Human, Mouse, Rat, Bovine, Xenopus tropicalis (Species predicted to react based on 100% sequence homology).

The protein encoded by this gene belongs to the protein phosphatase family, PP1 subfamily. PP1 is an ubiquitous serine/threonine phosphatase that regulates many cellular processes, including cell division. It is expressed in mammalian cells as three closely related isoforms, alpha, beta/delta and gamma, which have distinct localization patterns. This gene encodes the gamma isozyme. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PP1 gamma
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene PPP1CC
Gene Accession No. P36873
Gene Alias dis2m1; MGC1675; MGC2870; PP1; PP1C; PP1C gamma 1; PP1C gamma 2; PP1G; PP-1G; PP1gamma; PP2CG; PP2CGAMMA; PPP1CC; Ppp1cc1; PPP1G; PPP2CG; protein phosphatase 1 catalytic subunit gamma; protein phosphatase 1, catalytic subunit, gamma isoform; protein phosphatase 1, catalytic subunit, gamma isozyme; protein phosphatase 1C catalytic subunit; protein serine-threonine phosphatase catalytic subunit PP-1b; serine/threonine phosphatase 1 gamma; serine/threonine-protein phosphatase PP1-gamma catalytic subunit
Gene Symbols PPP1CC
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the center region of the Human PPP1CC.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5501
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.