Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PP11 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552934
Description
Immunogen sequence: CCKDFESLCS DHEVSHSSDA ITKEEIQSIS EKIYRADTNK AQKEDIVLNS QNCISPSETR NQVDRCPKPL FTYVNEKLFS KPTYAAFINL LNNYQRATGH GEHFSAQELA EQDAFLREIM KTAVMKELYS FLHHQNRY Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 83%.
This gene encodes a protein with protease activity and is expressed in the placenta. The protein may be useful as a tumor marker. Multiple alternatively spliced transcript variants have been found for this protein.
Specifications
PP11 | |
Polyclonal | |
Unconjugated | |
ENDOU | |
22 serine protease; 26 serine protease; endonuclease, poly(U) specific; endonuclease, poly(U)-specific; endonuclease, polyU-specific; ENDOU; P11; placental protein 11; placental protein 11 related; placental protein 11-related protein; Poly(U)-specific endoribonuclease; PP11; Pp11r; PP11-related protein; Protein endoU; PRSS26; T-cell-specific protein 30; Tcl-30; Uridylate-specific endoribonuclease | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8909 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P21128 | |
ENDOU | |
Recombinant protein corresponding to Human PP11. Recombinant protein control fragment (Product #RP-90729). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction