Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PP11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566924
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566924 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566924 Supplier Invitrogen™ Supplier No. PA566924
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KIESCASRCNE KFNRDAACQC DRRCLWHGNC CEDYEHLCTE DHKESEPLPQ LEEETEEALA SNLYSAPTSC QGRCYEAFDK HHQCHCNARC Q Highest antigen sequence identity to the following orthologs - mouse 64%, rat 64%.

This gene encodes a protein with protease activity and is expressed in the placenta. The protein may be useful as a tumor marker. Multiple alternatively spliced transcript variants have been found for this protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PP11
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ENDOU
Gene Accession No. P21128
Gene Alias 22 serine protease; 26 serine protease; endonuclease, poly(U) specific; endonuclease, poly(U)-specific; endonuclease, polyU-specific; ENDOU; P11; placental protein 11; placental protein 11 related; placental protein 11-related protein; Poly(U)-specific endoribonuclease; PP11; Pp11r; PP11-related protein; Protein endoU; PRSS26; T-cell-specific protein 30; Tcl-30; Uridylate-specific endoribonuclease
Gene Symbols ENDOU
Host Species Rabbit
Immunogen Recombinant Human PP11. Recombinant protein control fragment (Product #RP-105427).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8909
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.