Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PP11 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA580581
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human PP11.
This gene encodes a protein with protease activity and is expressed in the placenta. The protein may be useful as a tumor marker. Multiple alternatively spliced transcript variants have been found for this protein.
Specifications
PP11 | |
Polyclonal | |
Unconjugated | |
ENDOU | |
22 serine protease; 26 serine protease; endonuclease, poly(U) specific; endonuclease, poly(U)-specific; endonuclease, polyU-specific; ENDOU; P11; placental protein 11; placental protein 11 related; placental protein 11-related protein; Poly(U)-specific endoribonuclease; PP11; Pp11r; PP11-related protein; Protein endoU; PRSS26; T-cell-specific protein 30; Tcl-30; Uridylate-specific endoribonuclease | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
8909 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with no preservative | |
P21128 | |
ENDOU | |
A synthetic peptide corresponding to the center region of the Human PP11. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction