Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PP11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | PP11 |
---|---|
Concentration | 0.23 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-1928
![]() ![]() |
Proteintech Group Inc
180021AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This gene encodes a protein with protease activity and is expressed in the placenta. The protein may be useful as a tumor marker. Multiple alternatively spliced transcript variants have been found for this protein.Specifications
PP11 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
P21128, Q3V188 | |
19011, 680317, 8909 | |
PP11 Fusion Protein Ag12494 | |
Primary | |
-20°C | |
ENDOU |
0.23 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
26 serine protease, ENDOU, P11, Placental protein 11, PP11, Protein endoU, PRSS26 | |
Endou | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title