Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PP14/Glycodelin Antibody (SR1372), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | PP14/Glycodelin |
---|---|
Clone | SR1372 |
Dilution | Western Blot 1:500-1:2000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
PP14/Glycodelin Monoclonal antibody specifically detects PP14/Glycodelin in Human samples. It is validated for Western BlotSpecifications
PP14/Glycodelin | |
Western Blot 1:500-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
alpha uterine protein, GdF, GdS, glycodelin, glycodelin-A, glycodelin-F, glycodelin-S, MGC138509, MGC142288, PEP, Placental protein 14, PP14 protein (placental protein 14), PP14GDPAEGGdA, pregnancy-associated endometrial a, Pregnancy-associated endometrial alpha-2 globulin, pregnancy-associated endometrial alpha-2-globulin, progestagen-associated endometrial protein (placental protein 14, progestagen-associated endometrial proteinPEG, Progesterone-associated endometrial protein | |
A synthesized peptide derived from human PP14/Glycodelin (Uniprot #: P09466) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1372 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Developmental Biology | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
5047 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title